Burettes, Class B, with PTFE Detachable Stopcocks, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Made from high-quality accurate bore tubing for a high degree of accuracy, reliability and durability.
- Class B burette
- Graduated at intervals
- Available in multiple sizes
Detachable PTFE stopcock and tip facilitate easy cleaning and convenient replacement of broken tips.
Customers Who Bought This Also Bought