Science Take-Out® Nicotine Addiction: Breaking Up with My Vape
Fournisseur: SCIENCE TAKE-OUT, LLC
Certificats
A propos de cet article
Why is it so difficult to quit vaping?
- Available as a single kit with materials for 1 student or group
- Unassembled 10 pack for additional savings
- No additional equipment needed
Follow the story of a teen who is trying to quit vaping. Model how vaping nicotine affects the receptors and dopamine levels in the brain reward pathway. Use the model to understand nicotine tolerance, dependence, addiction, withdrawal and recovery from nicotine addiction.
Customers Who Bought This Also Bought