OneRack® Tube Racks, Polypropylene (PP)
Supplier: Heathrow Scientific
Certificates
About this item
OneRack® Tube Racks are tough and highly resistant to acids, bases, solvents, and heat.
- Made out of Polypropylene
- Have molded coordinates for tube orientation
- Stable design for use in water baths
- Flat labeling area for easier identification
- Unique tear drop shaped openings for better retention of tubes
- Offers good chemical and heat resistance (0 to 110 °C)
Durable OneRack® Tube Racks are molded in a single, continuous piece so no assembly is required and there are no detachable pieces.
Customers Who Bought This Also Bought