EZLabpure™ Media Bottle, Closed VersaCap, Sterile, Foxx Life Sciences
Fournisseur: Foxx Life Sciences
Certificats
A propos de cet article
The sterile EZlabpure™ Media Bottles, Polystyrene (PS) and PETG, have been designed specifically for Life Science researchers to store common solutions such as tissue culture media, serum, and buffers.
- PP white VersaCap
- Sloped shoulder for smooth pouring
- Molded finger grips
The lower height bottle has features that improve the media flow for easy pouring. The narrow bottle shoulder, combined with the molded finger grips, improves the ability to securely grip the bottle with gloved hands.
Certifications: Made with USP Class VI materials. Manufactured in a Class 100,000 cleanroom. Sterilized in the USA according to VDmax25 guidelines (SAL 10-6).
Customers Who Bought This Also Bought