Neuron Modeling
# de catalogue 470337-151
Fournisseur: 3-D Molecular Designs
Spécifications
- Description:Neuron modeling
- No. of Lab Groups:1
- Cat. no.:470337-151
Spécifications
A propos de cet article
A flexible neuron model
- Construct and describe a neuron
- Interactive modeling
- Reusable
Captivate your classroom and explore multiple concepts with our large and small foam neuron models!
Students can use the Neuron Modeling Kit© to:
Distinguish between multipolar neurons, bipolar neurons, unipolar neurons and interneurons, and determine their location and function in the human body.
Construct a model and identify parts of a multipolar neuron.
Use myelin sheath pieces to demonstrate differences between two types of neuroglia in the central and peripheral nervous systems.
Construct simple and complex neural pathways and examine the overall effect on neuronal firing at excitatory and inhibitory synapses.
Customers Who Bought This Also Bought