Making the Cut with CRISPR CAS-9
# de catalogue 470337-147
Fournisseur: 3-D Molecular Designs
Spécifications
- Description:Making the cut with CRISPR Cas-9
- No. of Lab Groups:1
- Cat. no.:470337-147
Spécifications
A propos de cet article
How is CRISPR used in biotechnology?
- Interactive modeling
- How does CAS-9 'Make The Cut'
- Reusable
Model how the CAS-9 protein is programmed with RNA to search for and identify specific sequences of DNA before making the double-stranded cut. Explore and discuss how CAS-9 is being engineered with new features that make it a useful tool for editing the human genome.
With making the cut with CRISPR-CAS9's colorful foam pieces and placemat your students can:
Build a schematic model of the CRISPR-CAS-9 RNA-guided endonuclease, including the viral double-stranded DNA and dual guide RNA.
Understand how the CAS-9 protein binds to a specific sequence of viral double-stranded DNA at a PAM site - and cuts it.
Demonstrate how that cut destroys viral genome and prevents the infection of bacteria.
And much more!
Customers Who Bought This Also Bought