Biotechnology Modeling Kit
# de catalogue 470337-145
Fournisseur: 3-D Molecular Designs
Spécifications
- Description:Biotechnology
- No. of Lab Groups:1
- Cat. no.:470337-145
Spécifications
A propos de cet article
Explore biotechnology topics
- Interactive modeling
- Covers PCR, restriction enzymes and Sanger sequencing
- Reusable
Help your students achieve a better understanding of multiple biotechnology concepts including diagnostics, forensics and bacterial transformation with a single hands-on modeling kit!
Using the color-coded foam nucleotides in the Biotechnology Kit© and teacher-developed activities, students:
Model polymerase chain reaction and how flanking primers result in the exponential amplification of a short product.
Model the Sanger DNA sequencing method using chain-terminating fluorescent dideoxy nucleotide analogs.
Model DNA cloning using restriction endonucleases to cut DNA at specific sequences.
And more!
Customers Who Bought This Also Bought