Flow of Genetic Information
# de catalogue 470337-143
Fournisseur: 3-D Molecular Designs
Spécifications
- Description:Flow of genetic information
- No. of Lab Groups:1
- Cat. no.:470337-143
Spécifications
A propos de cet article
How does DNA function?
- Interactive Modeling
- Replication , Transcription And Translation Modeling
- Reusable
Don't be surprised if one of your students blurts "Now I get it!" while using our Flow of Genetic Information Kit©. Manipulating the kit's color-coded foam nucleotides and placemats, students will model replication of the leading and lagging strands of DNA, model transcription as they copy one strand of DNA into mRNA using an RNA polymerase and model translation/protein synthesis as they decode the mRNA into protein on the ribosome placemat.
Customers Who Bought This Also Bought