Phospholipid and Membrane Transport
# de catalogue 470337-142
Fournisseur: 3-D Molecular Designs
Spécifications
- Description:Phospholipid membrane transport
- No. of Lab Groups:1
- Cat. no.:470337-142
Spécifications
A propos de cet article
How does a cell membrane operate?
- Interactive Modeling
- Explore Membrane Structure And Transport
- Reusable
Don’t tell your students about membranes and how/why they form – let them discover the spontaneous formation of membranes for themselves using this new field test kit featuring the amphipathic structure of phospholipids – with their hydrophilic heads and hydrophobic tails. Your students will explore the chemical structure of a phospholipid and then construct a phospholipid monolayer, a micelle and a bilayer leading to an understanding of the plasma membrane structure. Additional components of this kit allow students to construct lipid bilayers and consider the role of transport proteins in moving ions and small molecules across membranes.
Customers Who Bought This Also Bought