Autofil® 2 Bottle Top Filtration Devices, Hydrophilic PES Membrane, Sterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
Certificates
About this item
The Autofil® 2 features 0.1, 0.2, or 0.45 μm PES membranes. Perfect for filtering FBS and other viscous solutions. The membrane is low protein binding and surfactant-free.
- Funnel is compatible with GL 45 thread bottles
- Units include adapter for connecting to vacuum hose
- Design includes the NEW EZlabpure™ ergonomic bottle shape with molded finger grip
- GL 45 caps provides a convenient gripping surface with a frosted area for labeling
Autofil® 2 is available in 250, 500, or 1000 ml complete units with the NEW EZlabpure™ media bottle or funnel only bottle-top filters.
Certifications: Made with USP Class VI materials. Manufactured in a Class 100,000 cleanroom.
Frequently Bought Together