Reusable ESD Sheer V-Neck Labcoats, Short, ESD Knit Wrists, Uniform Technology
Fournisseur: Uniform Technology
Certificats
A propos de cet article
Reusable ESD labcoats are manufactured from 91% polyester/9% carbon.
- V-neck design
- Snap front
- 2 waist pockets, 1 chest pocket
- Snap wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.
Spécifications
- Catalog No:
- 76516-296
- 76516-294
- 76516-292
- 76516-290
- 76516-288
- 76516-286
- 76516-284
- 76516-282
- 76516-298
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 91% polyester/9% carbon
- 91% polyester/9% carbon
- 91% polyester/9% carbon
- 91% polyester/9% carbon
- 91% polyester/9% carbon
- 91% polyester/9% carbon
- 91% polyester/9% carbon
- 91% polyester/9% carbon
- 91% polyester/9% carbon