Reusable ESD StatStar Labcoats, Short, ESD Knit Wrists, Uniform Technology
Fournisseur: Uniform Technology
Certificats
A propos de cet article
Reusable ESD labcoats are manufactured from 87% polyester/13% carbon.
- Lapel collar
- Snap front
- 2 waist pockets, 1 chest pocket
- ESD knit wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.
Spécifications
- Catalog No:
- 76516-118
- 76516-116
- 76516-114
- 76516-112
- 76516-110
- 76516-108
- 76516-106
- 76516-104
- 76516-102
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD