PCRmax® Polypropylene PCR Microplates, Antylia Scientific
Supplier: Argos Technologies
Certificates
About this item
Ultra-thin and consistent wall thickness allows precise thermal transfer for optimal results.
- Manufactured with 99.9% pure virgin polypropylene
- RNAse/DNAse-free, nonpyrogenic, non sterile
- Compatible with sealing film
PCRmax® microplates are designed to fit real-time PCR thermal cyclers and sequencers. The construction of pure, clear polypropylene allows for full transparency. Microplates are compatible with PCRmax® Alpha Cycler instruments.
Customers Who Bought This Also Bought