Cole-Parmer® Essentials Jerrican with Handle, HDPE, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Better stability with a wider footprint.
- Integral spout allows accurate pouring
- Leak-proof screw closure
- Milky white polypropylene caps
- 1.53" (38.9 mm) neck I.D.
Rugged jerricans are built wide and low for better stability. Ideal for transporting and carrying a variety of liquids. Jerricans are not autoclavable.
Certifications: USP Class VI certified
Customers Who Bought This Also Bought