Cole-Parmer® Essentials Carboy with Handles, LDPE, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Integrated handles make transporting solutions easy and safe.
- Leakproof polypropylene cap
- Easy to read, molded graduations
- 5000 ml graduation intervals
These rugged, durable plastic carboys will meet large volume liquid storage and dispensing needs. Integrated wide handles make them easy to carry and pour, even with gloves on. Polypropylene cap with integrated seal ring ensures leakproof seal. The carboys maintain flexibility and shatterproof safety down to –103 °F ( –75 °C). Carboys are not autoclavable.
Customers Who Bought This Also Bought