Cole-Parmer® Essentials Carboy with Spigot, Rectangular, PP, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Maintain sample integrity.
- Leakproof for safe transport
- Autoclavable
These leakproof polypropylene rectangular carboys with spigot easily store and dispense sterile water and other solutions. Space-saving design requires less space than traditional round carboys. Features include built-in stainless steel handle and wide mouth opening for easy cleaning.
Certifications: USP Class VI certified
Customers Who Bought This Also Bought