Reusable ESD StatStar Labcoats, Long, Snap Wrists, Uniform Technology
Fournisseur: Uniform Technology
Certificats
A propos de cet article
Reusable ESD labcoats are manufactured from 87% polyester/13% carbon.
- Lapel collar
- Snap front
- 2 waist pockets, 1 chest pocket
- Snap wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.
Spécifications
- Catalog No:
- 76516-076
- 76516-074
- 76516-096
- 76516-094
- 76516-072
- 76516-098
- 76516-092
- 76516-070
- 76516-090
- 76516-068
- 76516-088
- 76516-066
- 76516-086
- 76516-084
- 76516-082
- 76516-080
- 76516-078
- 76516-100
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon
- 87% polyester/13% carbon