Cole-Parmer® Essentials Wide-Mouth Sample Polypropylene Containers, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Mix and store in one vessel.
- Easily attach to blenders for in-container mixing
- Durable for general storage—chemical-resistant polypropylene
- Autoclavable
Chemical-resistant polypropylene (PP) containers come with white PP linerless lids.
BodyTextAccessories: PP lid.
Customers Who Bought This Also Bought