VWR® Thermometer Case for Liquid-in-Glass Thermometers
# de catalogue 76491-684
Fournisseur: VWR International
Spécifications
- Cat. no.:76491-684
- For Use With:254 mm L thermometer
Spécifications
A propos de cet article
Open cases feature a screw cap with ring and small openings around bulb for circulation.
- Screw cap
- Ring for hanging
- Metal or nickel-plated brass
- Lower section surrounding the bulb with small openings for circulation
This case protects your thermometers from accidental breakage storage.
Customers Who Bought This Also Bought