VWR® Ultra Low Liquid-in-Glass Thermometers
Supplier: VWR International
Certificates
About this item
Partial and total immersion thermometers for low temperature applications.
- Include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
- Tested and inspected by our USA quality team
Designed for ultra-low temperature use to ensure that the meniscus will be visible in the appropriate low temperature applications, these thermometers feature mercury free liquid against lead-free, white back glass.
Certifications: Traceable to NIST. Compliant with ANSI/SAMA Z236.1-1983.
Customers Who Bought This Also Bought