VWR® Blood Bank Thermometer
# de catalogue 76491-642
Fournisseur: VWR International
Spécifications
- Accuracy:±1.0 °C
- Length:152 mm
- Cat. no.:76491-642
- Divisions:1.0 °C
- Range:−5...+20 °C
- Immersion:Total
Spécifications
A propos de cet article
This thermometer has been designed to indicate critical temperature range.
- Total immersion
- Individual serial numbers
- PTFE coating helps prevent liquid and glass contamination
The critical temperature range from 4 to 6 °C is marked with arrows.
Certifications: Traceable NIST. Envirokleen certified by ChemTel (an independent laboratory).
Customers Who Bought This Also Bought