VWR® Liquid-in-Glass Oven Thermometers
Supplier: VWR International
Certificates
About this item
These oven thermometers contain red organic liquid against white back glass.
- Partial immersion thermometers feature a round glass bulge in the body to hold them in place
- PTFE coating helps prevent liquid and glass contamination if the thermometer breaks
- Individually serialised
Customers Who Bought This Also Bought