VWR® Probeless Indoor/Outdoor Digital Thermometer
Catalog # 76491-652
Supplier: VWR International
Specifications
- Resolution:0.1 °C
- Protection class:Water Resistant IP65
- Description:Indoor/outdoor thermometer
- Cat. no.:76491-652
- Accuracy:±1.0 °C
- W×H:100×215 mm
- Range:−40...+50 °C
Specifications
About this item
Designed for use in high moisture or wet environments, this digital thermometer is used in green houses, sample prep areas, water treatment facilities and aquariums.
- Reset button allows for a new monitoring period
- Keyhole for mounting
- Easy to read
- Water resistant IP 65
Displays the current temperature and registers the maximum and minimum temperatures attained since the last reset.
Delivery: Includes LR44 battery.
Customers Who Bought This Also Bought