VWR® Calibrated Digital Thermometer/Event Logger with Stainless Steel Probe
Catalog # 76491-654
Supplier: VWR International
Specifications
- Resolution:0.1 °C
- Probe cable length:3 m (9.8')
- Cat. no.:76491-654
- W×H:65×110 mm
- Calibration Points:0, 100 °C
- Accuracy:±1.5 °C, ±0.5 °C (−20...+100 °C)
- Range:−50...+200 °C
Specifications
About this item
Monitor the temperature of pharmaceuticals, vaccines, chemicals, blood and blood products, food products and more with these high accuracy digital thermometers with chemical resistant probes.
- Probe length is 125 mm, stainelss steel
- 10-event recording with date/time stamp
- Max./min. temperature readings
- High/low alarm
- Calibration points: 0; 100 °C
This high accuracy digital thermometer has a stainless steel chemical resistant probe.
Delivery: Includes accredited calibration documentation and multi-language instructions (English, French, German, Italian, Portuguese and Spanish) and AAA battery.
Frequently Bought Together