VWR® Calibrated Digital Verification Thermometers, General Use
Supplier: VWR International
Certificates
About this item
Digital thermometers verify sample temperatures in freezers, refrigerators, and incubators.
- Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
- All models include accredited calibration documentation that meets the requirements of ISO 17025
- Tested and calibrated in the USA, traceable to NIST
Available calibrated for specific applications and equipment.
Certifications: ISO 17025, ISO/IEC 17025:2005, traceable to NIST.
Delivery: Includes multi-language instructions (English, French, German, Italian, Portuguese and Spanish), AAA or LR44 batteries.
Caution: Plastic bottles can not be used in ovens. Glass bottles and caps should not be used in conditions where temperatures may exceed 200 ˚C.
Customers Who Bought This Also Bought