VWR® Freezer and Ultra Low Freezer Verifictaion Thermometers
Supplier: VWR International
Certificates
About this item
These mercury free thermometers are traceable to NIST and cover ranges from -90 to 25 °C .
- Verify sample temperatures in freezers/ultra-low freezers
- Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
- Thermometers contain organic liquid, or EnviroKleen™ certified Enviro-Safe® liquid; feature permanently fused markings and lead free glass
- PFA coating safely contains glass and liquid in case of breakage
- Manufactured and tested by our USA quality team; individually serialized
Insert the thermometer into the bottle and place near the samples for best results.
Certifications: Traceable to NIST.
Delivery: Supplied with multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability plus VELCRO® and magnetic attachments.
Specifications
- Catalog No:
- 76491-670
- 76491-674
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health