United Scientific Supplies Dropper Bottle, Unassembled, LDPE, 15 ml
# de catalogue 470332-692
Fournisseur: United Scientific Supplies
Spécifications
- Capacity:15 ml
- Description:Dropper bottle, unassembled
- Material:LDPE
- Cat. no.:470332-692
Spécifications
A propos de cet article
This dropper bottle is made of translucent and low density polyethylene.
- Bottle can be used with or without dropper
- Comes in a pack of 24
For use when dispensing small drops of a solution. The drop sizes are uniform for consistent dispensing of liquids.
Customers Who Bought This Also Bought