ContactUsLinkComponent
United Scientific Supplies Dropper Bottle, Assembled, LDPE, 30 ml
United Scientific Supplies Dropper Bottle, Assembled, LDPE, 30 ml
# de catalogue 470332-794
DROPPER BOTTLE, ASSEMBLED, LDPE, 30ML
United Scientific Supplies Dropper Bottle, Assembled, LDPE, 30 ml
# de catalogue 470332-794
Spécifications
  • Capacity:
    30 ml
  • Description:
    Dropper bottle, assembled
  • Material:
    LDPE
  • Cat. no.:
    470332-794
Spécifications
A propos de cet article
This dropper bottle is made of translucent and low density polyethylene.
  • For use when dispensing small drops of a solution
  • Drop sizes are uniform for consistent dispensing of liquids
Bottle can be used with or without dropper.
Customers Who Bought This Also Bought