United Scientific Supplies Dropper Bottle, Assembled, LDPE, 30 ml
# de catalogue 470332-794
Fournisseur: United Scientific Supplies
Spécifications
- Capacity:30 ml
- Description:Dropper bottle, assembled
- Material:LDPE
- Cat. no.:470332-794
Spécifications
A propos de cet article
This dropper bottle is made of translucent and low density polyethylene.
- For use when dispensing small drops of a solution
- Drop sizes are uniform for consistent dispensing of liquids
Bottle can be used with or without dropper.
Customers Who Bought This Also Bought