United Scientific Supplies Specimen Jars, Screw-Cap, Glass
Fournisseur: GSC International
Certificats
A propos de cet article
Flint glass specimen jars for easy storage.
- Autoclavable
All the wide mouth jars are made from soda lime glass and have a PP cap with a PE liner. Since the opening is so large, it is easy to insert and remove materials.
Customers Who Bought This Also Bought