Reusable ESD Sheer Labcoats, Short, ESD Knit Wrists, Uniform Technology
Fournisseur: Uniform Technology
Certificats
A propos de cet article
Reusable ESD labcoats are manufactured from 91% polyester/9% carbon.
- Lapel collar
- Snap front
- 2 waist pockets, 1 chest pocket
- ESD knit wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.
Spécifications
- Catalog No:
- 76516-276
- 76516-274
- 76516-272
- 76516-270
- 76516-268
- 76516-266
- 76516-264
- 76516-280
- 76516-278
- Model:
- BR3C-42
- BR3C-42
- BR3C-42
- BR3C-42
- BR3C-42
- BR3C-42
- BR3C-42
- BR3C-42
- BR3C-42
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 91% polyester/9% carbon
- 91% polyester/9% carbon
- 91% polyester/9% carbon
- 91% polyester/9% carbon
- 91% polyester/9% carbon
- 91% polyester/9% carbon
- 91% polyester/9% carbon
- 91% polyester/9% carbon
- 91% polyester/9% carbon