Reusable Altessa Grid Cleanroom Frocks with Zipper Closure, Uniform Technology
Fournisseur: Uniform Technology
Certificats
A propos de cet article
Cleanroom frocks are manufactured from 99% polyester/1% carbon fabric.
- Military collar
- Raglan sleeves
- ESD knit wrists
- Coil zipper with top snaps
Suitable for ISO Class 5 or higher cleanrooms.
Spécifications
- Catalog No:
- 76515-724
- 76515-722
- 76515-720
- 76515-718
- 76515-696
- 76515-694
- 76515-692
- 76515-736
- 76515-690
- 76515-734
- 76515-732
- 76515-688
- 76515-730
- 76515-686
- 76515-728
- 76515-684
- 76515-726
- 76515-702
- 76515-700
- 76515-698
- 76515-716
- 76515-714
- 76515-712
- 76515-710
- 76515-708
- 76515-706
- 76515-704
- Model:
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- CFRZC-74
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon
- 99% polyester/1% carbon