Vikan® Long Handle, Narrow Head Cleaning Brushes, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
Easily clean small spaces with this narrow-headed cleaning brush.
- Polyester bristles, polypropylene block
The bristles are angled to allow easy cleaning of corners, the grip is designed for extra stability and the entire brush is color-coded to support HACCP principles.
Overall dimensions: 16.54L×1.81W×4.33"H
Customers Who Bought This Also Bought