Vikan® UST Deck Scrub Brushes, Stiff Bristles, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
UST 16" deck scrub brushes with stiff bristles.
- Polyester bristles, polypropylene block
- Euro (hygienic) thread type
FDA compliant.
Overall dimensions: 15.25L×1.88W×3.9"H
Customers Who Bought This Also Bought