Vikan® Deck/Wall Scrub Brushes, Stiff Bristles, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
This floor scrubbing brush features firm bristles.
- Polyester bristles, polypropylene block
- Euro (hygienic) thread type
The bristle length is perfect for scrubbing all types of floors, epoxy, tiles, cement, etc. It also reaches well into holes and grouting to remove hidden dirt.
Overall dimensions: 11.8L×2.8W×4.5"H
Customers Who Bought This Also Bought