Vikan® High-Low Brushes, Medium Bristles, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
The angled shape of this scrubbing brush is designed to clean the area where the wall meets the floor, but it is also effective for cleaning the floor itself.
- Polyester bristles, polypropylene block
- Euro (hygienic) thread type
The bristles are gentle enough to be used on equipment. It is a popular choice in meat processing facilities, canneries, mills, wineries and breweries and generally any moist environment that creates difficult cleaning challenges.
Overall dimensions: 10.4L×5.5W×3.9"H
Customers Who Bought This Also Bought