Vikan® Compact Deck Scrubs, Stiff Bristles, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
Clean around equipment and against walls with this HACCP compliant color-coded deck/wall scrub.
- Polyester bristles, polypropylene block
- Euro (hygienic) thread type
This scrub brush features angled bristles around the perimeter and a rounded, compact design for improved durability.
Overall dimensions: 8.86L×2.95W×4.53"H
Customers Who Bought This Also Bought