Vikan® Grout Brushes, Extra Stiff Bristles, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
Excellent for scrubbing in difficult to reach areas or areas with limited space.
- Polyester bristles, polypropylene block
- Euro (hygienic) thread type
The handle can be rotated down so that it is parallel to the floor, providing access underneath equipment that is as low as 5" from the floor. The bristles are cut in an 'arrow' shape to facilitate the cleaning of grout between tiles and crevices in general. The middle row of bristles are stiffer than the filament bundles on the sides. The new crevice scrub is mounted with a thread adaptor that can be removed for easy cleaning of the whole product.
Overall dimensions: 9.25L×1.5W×6.25"H
Customers Who Bought This Also Bought