Vikan® Tank Brushes, Stiff Bristles, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
This oval-shaped brush with stiff bristles is effective for scrubbing inside large industrial tanks and other large container-like equipment.
- Polyester bristles, polypropylene block
- Euro (hygienic) thread type
The rounded edges work well to clean down the sides of equipment. It also fits well into large channel drains.
Overall dimensions: 8.3L× 5.1W×3.7"H
Customers Who Bought This Also Bought