Vikan® Small Hand Scrapers, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
This color-coded small hand scraper is an excellent solution for all bench top scraping needs.
- Polypropylene construction
- Seamless design
- Length: 7.8"
It has no seams or cracks, which helps prevent bacterial growth.
Customers Who Bought This Also Bought