Vikan® Upright Dust Pans, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
The contact lip is designed for easy and efficient sweeping into the dustpan.
- Polypropylene pan and grip
- Aluminum handle
The outer front surface has a raised area to prevent debris from fallling out of the dustpan. The enlarged rear and side walls enable the dustpan to hold larger volumes of debris.
Overall dimensions: 12.9L×7.87W×33.86"H
Customers Who Bought This Also Bought