Vikan® Bench Brushes, Medium Bristles, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
This is a dusting brush with a smooth, ergonomically designed handle. The medium strength bristles remove moist or medium sized particles, such as sugar, chocolate, pastry, or vegetable peelings.
- Color-coded to support HACCP principles
- Polypropylene block, polyester bristles
Also suitable for use as a dustpan brush on floors.
Overall dimensions: 13L×1.3W×3.3"H
Customers Who Bought This Also Bought