Vikan® Baker's Brushes, Soft Bristles, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
This long, narrow,fully color-coded hand brush is perfect for dusting large surface areas.
- Color-coded to support HACCP principles
- Polypropylene block, polyester bristles
Soft bristles are designed for brushing dry, fine particles like dust, flour, and powdered sugar.
Overall dimensions: 11L×1.2W×3.7"H
Customers Who Bought This Also Bought