Vikan® Detail Brushes, Extra Stiff Bristles, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
Small compact hand brush suitable for cleaning in and around gaskets, rubber strips, and joints on machinery.
- Color-coded to support HACCP principles
- Polypropylene block, polyester bristles
Ideal for cleaning equipment link conveyor belts.
Overall dimensions: 7.9L×0.5W×0.9"H
Customers Who Bought This Also Bought