Vikan® High-Temp Hand Brush, Remco
# de catalogue 76528-978
Fournisseur: Remco Products
Spécifications
- Description:High-temp hand brush, white
- Cat. no.:76528-978
Spécifications
A propos de cet article
Originally designed for removing debris from heat sealers and other packing equipment, this brush is also ideal for cleaning hot grills and frying pans.
- Clean safely without contacting hot surfaces
- Polypropylene block, polyester bristles
Ergonomically friendly design with a long handle for safe cleaning.
Overall dimensions: 11.42L×1.18W×2.56"H
Customers Who Bought This Also Bought