ContactUsLinkComponent
Vikan® High-Temp Hand Brush, Remco
Vikan® High-Temp Hand Brush, Remco
# de catalogue 76528-978
Fournisseur:  Remco Products
Brush Heat Resist Stiff 11.5in Wht, With an ergonomic short handle and very soft bristles that retain water and chemcials to improve cleaning efficacy, washing brush is perfect for cleaning fragile surfaces such as glass.
Vikan® High-Temp Hand Brush, Remco
# de catalogue 76528-978
Fournisseur:  Remco Products
Spécifications
  • Description:
    High-temp hand brush, white
  • Cat. no.:
    76528-978
Spécifications
A propos de cet article
Originally designed for removing debris from heat sealers and other packing equipment, this brush is also ideal for cleaning hot grills and frying pans.
  • Clean safely without contacting hot surfaces
  • Polypropylene block, polyester bristles
Ergonomically friendly design with a long handle for safe cleaning.
Overall dimensions: 11.42L×1.18W×2.56"H
Customers Who Bought This Also Bought