Vikan® Short Handle Washing Brushes, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
Ergonomic short handle with very soft bristles that retain water and chemicals for cleaning efficancy.
- Color-coded to support HACCP principles
- Polypropylene block, polyester bristles
Use to clean fragile surfaces such as glass. Also suitable for coating.
Overall dimensions: 10.63L×2.76W×3.54"H
Customers Who Bought This Also Bought