Vikan® Short Handle Scrubbing Brushes, Stiff Bristles, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
This fully color-coded, short-handled wash brush has a comfortable grip, allowing extended work without hand fatigue.
- Color-coded to support HACCP principles
- Polypropylene block, polyester bristles
The stiff bristles are great for washing all types of equipment, tables, and walls. Use the brush holding both the handle and the brush head for better control.
Overall dimensions: 9.8L×2.6W×2.6"H
Customers Who Bought This Also Bought