Vikan® Narrow Long-Handle Cleaning Brushes, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
Easily clean small space with this narrow head brush.
- Color-coded to support HACCP principles
- Polypropylene block, polyester bristles
The bristles are angled to facilitate easy cleaning of corners. The grip is designed for extra stability and the entire brush is color-coded.
Overall dimensions: 16.54L×1.81W×4.33"H
Customers Who Bought This Also Bought