Vikan® UST Short Handle Scrubbing Brushes, Stiff Bristles, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
This UST short handled hand brush with angled Bristle Security Units is effective when cleaning conveyor belts, production lines, machinery, and food preparation surfaces in high risk areas.
- Polypropylene block, polyester bristles
Angled and circular arrangement of Bristle Security Units ensures effcient cleaning.
Customers Who Bought This Also Bought