Vikan® Flexible Hand Scrapers, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
Flexible scraper is suitable for cleaning work benches and conveyor belts, as well as empyting containters and buckets.
- Equipped with a scraping edge on 3 sides
- Polypropylene construction
Scraper dimensions: 6.5L×0.08W×3.62"H
Customers Who Bought This Also Bought