Vikan® Handle Mounted Stainless Steel Scraper, Remco
Fournisseur: Remco Products
Certificats
A propos de cet article
Scrapers feature a color-coded, threaded grip that allows cleaning of difficult to reach areas.
- Suitable for removing stubborn debris
- Stainless steel blade, polypropylene grip
- Euro (hygienic) thread type
- Available in 2" or 4" blade widths
Slightly rounded blade corners prevent damage to surfaces, and reduce operator injury.
Customers Who Bought This Also Bought